Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium pentaborate octahydrate ≥99%

Supplier: BeanTown Chemical

223240-2KG 223240-500G
BT223240-500GEA 62.42 USD InStock
BT223240-500G BT223240-2KG
Ammonium pentaborate octahydrate ≥99%
Ammonium pentaborate octahydrate
  • Powder


Formula: H₄B₅NO₈·8H₂O
MW: 344.21 g/mol
MDL Number: MFCD00135839
CAS Number: 12046-03-6
EINECS: 234-521-1
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty