Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylsulfonium iodide ≥99%

Supplier: BeanTown Chemical

224470-1KG 224470-250G 224470-50G
BT224470-250GEA 213.96 USD InStock
BT224470-250G BT224470-50G BT224470-1KG
Trimethylsulfonium iodide ≥99%
Trimethylsulphonium iodide
  • RTECS: WR8750000
  • Light Sensitive
  • Crystalline


Formula: C₃H₉IS
MW: 204.07 g/mol
Melting Pt: 215 °C (subl.)
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty