Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultramark® 1621

Supplier: BeanTown Chemical
Synonyms: Massmark 1621

225160-250MG 225160-1G
BT225160-1GEA 165.98 USD InStock
BT225160-1G BT225160-250MG
Ultramark® 1621
Ultramark® 1621
  • Refractive index: 1.353
  • Liquid


Formula: C₆H₁₈N₃P₃
MW: 231.189 g/mol
Density: 1.8 g/mL
MDL Number: MFCD00242454
CAS Number: 105809-15-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty