Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-α-Phenylalanine ≥99%

Supplier: BeanTown Chemical
Synonyms: 3-Phenyl-D(+)-alanine3-Phenyl-D-alanineH-D-Phe-OHD-Phenylalanine

225200-100G 225200-25G 225200-5G
BT225200-25GEA 100.76 USD InStock
BT225200-25G BT225200-100G BT225200-5G
D-α-Phenylalanine ≥99%
D-α-Phenylalanine
  • RTECS: AY7533000
  • Powder


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00004270
CAS Number: 673-06-3
EINECS: 211-603-5
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty