Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubidium acetate ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

225455-10G
BT225455-10GEA 67.73 USD InStock
BT225455-10G
Rubidium acetate ≥99.5% (trace metals basis)
Rubidium acetate
  • Moisture Sensitive
  • Crystalline


Formula: H₃CCOORb
MW: 144.51 g/mol
Melting Pt: 246 °C
MDL Number: MFCD00013058
CAS Number: 563-67-7
EINECS: 209-255-4
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty