Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 4,000

Supplier: BeanTown Chemical
Synonyms: PEG 4,000PEG 4000

225900-250G 225900-1KG 225900-5KG
BT225900-1KGEA 117.94 USD InStock
BT225900-1KG BT225900-250G BT225900-5KG
Polyethylene glycol 4,000
Polyethylene glycol 4,000
  • RTECS: TQ4050000
  • Powder


Formula: H(OCH₂CH₂)nOH
MW: 4000 g/mol
Melting Pt: 58…62 °C
Density: 1.212 g/mL
MDL Number: MFCD01779612
CAS Number: 25322-68-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty