Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alpha 3 Rocket

Supplier: ESTES INDUSTRIES INC SE
1256
470006-855CS 711.5 USD InStock
470006-855
Alpha 3 Rocket
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Join the hundreds of thousands of people who got their start in model rockets with this classic!

  • Length: 12.3" (31.2 cm)
  • Diameter: .98" (25 mm)
  • Weight: 1.2 oz. (34 g)
  • Recommended Engines (not included): 1/2A6-2, A8-3 (First Flight), B4-4, B6-4, B6-6, C6-5, C6-7
  • Projected Max Altitude: 1100 ft. (335 m)

The Alpha III was the very first rocket in the E2X - Easy to Assemble series. This rocket series was designed with the first time rocketeer in mind. With all precolored rocket parts and easily applied self-stick decals, you're ready for liftoff in minutes. The black body tube, fluorescent orange nose cone and one piece fin unit quickly builds into one flashy rocket!

With nothing more needed than glue, you can be on the flying field in no time at all! Using standard engines, the Alpha III soars to heights over 1,100 feet. Whether you're a first time rocketeer or a long time flier, this 12 inch rocket is a true classic that should be in every collection. E2X - Easy to Assemble. Plastic Fins, Self-stick Decals, 12 in. (30.5 cm) Parachute Recovery.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty