Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgY Goat Antibody (CF™ 594)

Supplier: Biotium

20114-1MG
89493-212EA 256.16 USD InStock
89493-212
Anti-IgY Goat Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgY
Clonality:
Clone:
Conjugation: CF™ 594
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty