Would you like to log in again?
PCR Markers, 50 to 2000 bp, MilliporeSigma
Supplier: MilliporeSigma
69278-3 69278-3
80030-996EA 237.04 USD InStock
80030-996 CA80030-996
PCR Markers, 50 to 2000 bp, MilliporeSigma
Electrophoresis Reagents Molecular Weight Markers
Mixture of eight defined DNA fragments for characterizing small DNA products.
  • Ready to use
  • DNA fragments ranging from 50 to 2000 bp
  • Two tracking dyes
  • Easy to photograph
The markers are supplied ready-to-use in gel loading buffer containing two tracking dyes that do not interfere with UV illumination of ethidium bromide-stained bands. Each vial of markers is enough for 50 lanes on TBE or TAE agarose gels stained with ethidium bromide. 5 µL produces bands of even intensity that are bright, sharp, and easy to photograph.
Delivery information: A separate vial of 6X Loading Buffer is included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty