Would you like to log in again?
Screw Caps for Micro Tubes, Sarstedt
Supplier: Sarstedt
65.716.306 65.716.308 65.716.399 65.716.307 65.716.305 65.716.300 65.716.310 65.716.003 65.716.304 65.716.309
101093-841PK 63.08 USD InStock
101093-841 101093-831 101093-843 101093-833 101093-823 101093-835 101093-736 101093-837 101093-839 101093-829
Screw Caps for Micro Tubes, Sarstedt
Closures Test/Sample Tube Closures
All screw caps are made from polypropylene and feature an integral O-Ring
  • Polypropylene construction
  • Colored screw caps for easy identification
Caps provide maximum safety for storage, transport, and centrifugation.
Ordering information: The assorted color pack contains a mix of 10 colors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty