Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Semi-Micro Cuvette, Sarstedt
Supplier: Sarstedt
67.742
101093-934PK 21.77 USD InStock
101093-934
Semi-Micro Cuvette, Sarstedt
Cuvettes Spectrophotometer Cuvettes
Semi-micro polystyrene cuvettes with two optical sides
Suitable for use in 330nm wavelength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty