Would you like to log in again?
Pierce™ Protease Inhibitor Tablets and Mini Tablets, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™
A32961 A32959 A32953 A32955 A32965 A32963
PIA32959EA 368.4 USD InStock
PIA32959 PIA32955 PIA32953 PIA32965 PIA32963 PIA32961
Pierce™ Protease Inhibitor Tablets and Mini Tablets, Thermo Scientific
Protein Purification Protein Purification Kits
The Pierce™ protease and phosphatase inhibitor tablets contains protease and/or phosphatase inhibitors to provide broad spectrum complete protections to proteins from degradations.
  • The inhibitor tablests form quickly dissolves to give clear formulation
  • EDTA inhibits metalloproteases by chelating divalent cations necessary for their activity
  • The inhibitor formulation is compatible with most protein assays
  • Do not contain phosphatase inhibitors
  • Ready-to-use and fully disclosed
  • Can be used directly with Pierce BCA assays
Each tablet contains a mixture of several potent inhibitors. The protease inhibitors target aminopeptidases and cysteine and serine proteases. The phosphatase inhibitors target serine, threonine and protein tyrosine phosphatases.
Protease inhibitor tablets protect proteins from proteolytic degradation through inhibition of the serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin, and pepstatin A.
Caution: Store at 2°C to 8°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty