Would you like to log in again?
Wort Broth (Base), MilliporeSigma
Supplier: MilliporeSigma
1.05449.0500
EM1.05449.0500EA 165.76 USD InStock
EM1.05449.0500
Wort Broth (Base), MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
  • Good to very good growth in Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)
  • Good to very good growth in Saccharomyces cerevisiae ATCC 9080
  • Good to very good growth in Geotrichum candidum DSM 1240
  • Good to very good growth in Rhodotorula mucilaginosa DSM 70403
  • Good to very good growth in Penicillium commune ATCC 10428
  • Good to very good growth in Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)
  • Good to very good growth in Trichophyton ajelloi ATCC 28454
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty