Would you like to log in again?
Sucrose, MilliporeSigma
Supplier: MilliporeSigma
1.07651.5000
EM1.07651.5000EA 334.73 USD InStock
EM1.07651.5000
Sucrose, MilliporeSigma
Media for Microbiology Additives and Supplements for Microbiology
  • LD 50 oral, LD50 Rat 29700 mg/kg
  • Passes test for IR-spectrum
  • Less than or equal to 0.001% heavy metals as Pb
  • Less than or equal to 0.5% loss on drying at 105°C
  • 66.2 to 66.8% Spec. rotation (α 20/D; 26 %; water)
Suitable for microbiology and TLC.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty