Would you like to log in again?
GENPack for KidWind
Supplier: VERNIER SOFTWARE & TECH SE
KW-GP
470165-144EA 78.99 USD InStock
470165-144
GENPack for KidWind
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
The GENPack allows students to construct their own electrical generator and perform experiments with electricity and magnetism. Changing variables in the generator design will impact current and voltage output. The GENPack is perfect for physics and electrical students exploring Faraday’s law, induction, AC/DC electricity production, and energy transformations. We recommend this kit for advanced middle school students and older.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty