Would you like to log in again?
High Torque Generator with Wires
Supplier: VERNIER SOFTWARE & TECH SE
KW-HIGEN
470165-146EA 8.99 USD InStock
470165-146
High Torque Generator with Wires
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
The High Torque Generator is another option for your wind turbine generator. When compared to the standard Wind Turbine Generator, this High Torque Generator will produce lower voltage but higher current for a given rotor RPM. The High Torque Generator is great for load devices with higher current draw, such as the water pump.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty