Would you like to log in again?
KidWind Nacelle
Supplier: VERNIER SOFTWARE & TECH SE
KW-NAC
470165-150EA 17.99 USD InStock
470165-150
KidWind Nacelle
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Build a complete turbine by making your own tower and base with PVC pipe (from a hardware store) or use the KidWind Tower and Base Set. You will also need a generator and a way to affix the turbine blades. We recommend using the KidWind Wind Turbine Generator with Wires and a KidWind Hub.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty