Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Annexin V-Cy3 Apoptosis Kit, BioVision

Supplier: BioVision
K102-400 AB14142-100 K102-25
10186-384EA 537.74 USD InStock
10186-384 10186-386 10186-388
Annexin V-Cy3 Apoptosis Kit, BioVision
Assays Cellular Assays
Detects Apoptosis within 10 min.

Detection method- Flow cytometry (Ex = 543 nm; Em = 570 nm) and fluorescence microscopy
Species reactivity- Mammalian
Kit size- Convenient sizes (100 assays)

Annexin V Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, cells translocate the membrane phosphatidyl-serine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can be easily detected by staining with a fluorescent conjugate of Annexin V, a protein that has a high affinity for PS. The one-step staining procedure takes only 10 minutes. Detection can be analyzed by flow cytometry or by fluorescence microscopy.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty