Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Annexin V-Biotin Apoptosis Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB273267-25TEST
10186-414EA 306.43 USD InStock
10186-414
Annexin V-Biotin Apoptosis Kit, BioVision
Assays Cellular Assays
Detects Apoptosis by FACS or FL

Detection method- Flow cytometry (Ex = 488 nm; Em = 530 nm) using and fluorescence microscopy
Species reactivity- Mammalian
Kit size- Convenient sizes (25, 100, 400 assays)
Applications- Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis.

The Annexin V-Biotin Apoptosis Detection Kit is based on the observation that soon after initiating apoptosis, most cell types translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can easily bind to Biotin-conjugated Annexin V, a protein that has a strong natural affinity for PS. Annexin V-Biotin can be detected in conjunction with conventional dye-staining using any streptavidine- or avidin-dye reagents, such as (strept)avidine-fluorescein, -peroxidase, -alkaline phosphatase (AP), and -β-gal, etc.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty