Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caspase-1 Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB273268-25TEST
10186-422EA 306.43 USD InStock
10186-422
Caspase-1 Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Convenient, Highly-Sensitive Assay

Detection method- Absorbance (400- or 405 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (25, 100, 200, 400 assays)
Applications- Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis.

Activation of ICE-family proteases/caspases initiates apoptosis or other cellular processes in mammalian cells. The Caspase-1/ICE Colorimetric Protease Assay Kits provide a simple and convenient means for assaying the activity of caspases that recognize the sequence YVAD. The assay is based on spectrophotometric detection of the chromophore p-nitroanilide (pNA) after cleavage from the labeled substrate YVAD-pNA. The pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405 nm. Comparison of the absorbance of pNA from a treated sample with an untreated control allows determination of the fold increase in Caspase-1 activity.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty