Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caspase-4 Fluorometric Assay Kit, BioVision

Supplier: BioVision
K126-200 AB65658-100 K126-25 K126-400
10186-442EA 939.83 USD InStock
10186-442 10186-432 10186-434 10186-436
Caspase-4 Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Convenient, Highly-Sensitive Assay

Detection method- Fluorescence (Ex/Em 400/505 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (25, 100, 200, 400 assays)
Applications- Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis.

Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. The Caspase-4 Fluorometric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence LEVD. The assay is based on detection of cleavage of substrate LEVD-AFC (AFC: 7-amino-4-trifluoromethyl coumarin). LEVD-AFC emits blue light (400 nm); upon cleavage of the substrate by caspase-4 or related caspases, free AFC emits a yellow-green fluorescence (505 nm), which can be quantified using a fluorometer or a fluorecence microtiter plate reader.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty