Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Enhanced Apoptotic DNA Ladder Detection Kit, BioVision

Supplier: BioVision
AB66093-50
10186-484EA 472.15 USD InStock
10186-484
Enhanced Apoptotic DNA Ladder Detection Kit, BioVision
Assays Cellular Assays
Detects Apoptotic DNA within 90 min., Highly Sensitive

Detection method- Agarose gel electrophoresis
Species reactivity- Mammalian
Kit size- 50 assays
Applications- Detects efficiently DNA fragmentation from apoptotic cells

Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells. BioVision’s Enhanced Apoptotic DNA Ladder Detection Kit provides an easy and sensitive means for detecting DNA fragmentation in apoptotic cells. Unlike other commercially available kits that require 1-2 days to perform the procedure, the new detection method requires less than 90 minutes to prepare DNA, with neither extraction nor using columns. DNA fragmentation can be easily visualized by agarose gel electrophoreses stained with a highly sensitive dye.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty