Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cathepsin B Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65300-100
10186-486EA 609.17 USD InStock
10186-486
Cathepsin B Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Fluorescence (Ex/Em 400/505) and fluorometer
Species reactivity- Mammalian
Kit size- 100 assays
Applications- Detects alternative proteolytic enzymes such as the lysosomal cathepsin proteases that can initiate or propagate proapoptotic signals.

Apoptosis can be mediated by mechanisms other than the traditional caspase-mediated cleavage cascade. There is growing recognition that alternative proteolytic enzymes such as the lysosomal cathepsin proteases may initiate or propagate proapoptotic signals. Cathepsins are lysosomal enzymes that are also used as sensitive markers in various toxicological investigations. The Cathepsin-B Activity Assay kit is a fluorescence-based assay that utilizes the preferred cathepsin-B substrate sequence RR labeled with AFC (amino-4-trifluoromethyl coumarin). Cell lysates or other samples that contain cathepsin-B will cleave the synthetic substrate RR-AFC to release free AFC. The released AFC can easily be quantified using a fluorometer or fluorescence plate reader.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty