Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cathepsin S Inhibitor Screening Kit, BioVision (Fluorometric)

Supplier: BioVision
AB185437-100TEST
10186-492EA 942.29 USD InStock
10186-492
Cathepsin S Inhibitor Screening Kit, BioVision (Fluorometric)
Assays Cellular Assays
Only kit on the market to screen Cathepsin S inhibitors

Detection method- Fluorescence (Ex/Em = 400/505 nm)
Applications- Screen/study/characterize potential inhibitors of Cathepsin S.

Cathepsin S (CTSS, EC 3.4.22.27) is a lysosomal cysteine proteinase that is suggested to participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. Biovision’s Cathepsin S Inhibitor Screening Kit utilizes the ability of Cathepsin S to cleave the synthetic AFC based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a Cathepsin S inhibitor, the cleavage of the substrate is reduced/abolished resulting in decrease or total loss of the AFC fluorescence. This high-throughput adaptable assay kit is simple, sensitive, and rapid tool to screen the potential inhibitors of Cathepsin S.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty