Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Granzyme B Inhibitor Screening Kit, BioVision (Fluorometric)

Supplier: BioVision
AB157405-100TEST
10186-498EA 1179.78 USD InStock
10186-498
Granzyme B Inhibitor Screening Kit, BioVision (Fluorometric)
Assays Cellular Assays
Simple Assay, HTS

Detection method- Fluorescence ( Ex/Em = 380/500 nm )
Applications- Screening Granzyme B inhibitors, characterize/Study Granzyme B inhibitors.

Granzyme B (GZMB, EC number 3.4.21.79) also known as Granzyme-2 is a serine protease stored in the granules of activated cytotoxic T cells and NK cells. Upon target cell contact, Granzyme B is directionally exocytosed and with the assistance of perforin enters the target cell. With its unique substrate specificity (Granzyme B prefers an aspartic acid residue at the P1 site of its substrates), Granzyme B processes and activates various pro-caspases thereby inducing apoptosis in the target cell. In BioVision’s Granzyme B Inhibitor Screening Kit, active human Granzyme B enzyme hydrolyzes the specific substrate to release the quench of fluorescent group, which can be detected fluorometrically at Ex/Em = 380/500 nm. In the presence of potent Granzyme B Inhibitor, the hydrolyzation of substrate will be impeded. The Kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput screening of Granzyme B inhibitors or characterize/study Granzyme B inhibitors. Inhibitor control, Ac-IEPD-CHO is included to compare the efficacy of test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty