Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CaspGLOW™ Fluorescein Active Caspase Staining Kit, BioVision

Supplier: BioVision
AB65611-100
10186-502EA 603.3 USD InStock
10186-502
CaspGLOW™ Fluorescein Active Caspase Staining Kit, BioVision
Assays Cellular Assays
Detects activated Caspases by FACS, FL or plate reader

Detection method- Fluorescence microscopy, Flow cytometry and fluorescence plate reader (Ex. = 485 nm and Em. = 535 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (25 and 100 assays)
Applications- Sensitive detection of activated caspases in living cells.

Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Active Caspase Staining Kit provides a convenient and sensitive means for detecting activated caspases in living cells. The assay utilizes the caspase family inhibitor VAD-FMK conjugated to FITC (FITC-VAD-FMK) as a marker. FITC-VAD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspases in apoptotic cells.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty