Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CaspGLOWTM Red Active Caspase-9 Staining Kit, BioVision

Supplier: BioVision
AB65619-100
10186-526EA 563.96 USD InStock
10186-526
CaspGLOWTM Red Active Caspase-9 Staining Kit, BioVision
Assays Cellular Assays
Detects activated Caspase-9 by FACS, FL or plate reader

Detection method- Fluorescence microscopy, fluorescence plate reader (Ex/Em = 540/570 nm) and Flow cytometry
Species reactivity- Mammalian
Kit size- Convenient sizes (25 and 100 assays)
Applications- Detection of activated caspase-9 in living cells.

Activation of caspases plays a central role in apoptosis. The CaspGLOWTM Red Active Caspase-9 Staining Kit provides a convenient means for detecting activated caspase-9 in living cells. The assay utilizes a caspase-9 inhibitor LEHD-FMK conjugated to sulfo-rhodamine (Red-LEHD-FMK) as the fluorescent marker. Red-LEHD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-9 in apoptotic cells.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty