Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Annexin V-Cy3 Apoptosis Kit Plus, BioVision

Supplier: BioVision
AB14144-100
10186-540EA 531.17 USD InStock
10186-540
Annexin V-Cy3 Apoptosis Kit Plus, BioVision
Assays Cellular Assays
Detects Apoptotic, Necrotic & Healthy cells within 10 min

Detection method- flow cytometry using FL1 channel for SYTOX Green dye (Ex = 488 nm; Em = 530 nm) and FL2 channel for Annexin V-Cy3 (Ex = 543 nm; Em = 570 nm).
Species reactivity- Mammalian
Kit size- Convenient sizes (25, 100 and 400 assays)
Applications- Detecting apoptosis in living cells by flow cytometry or fluorescence microscopy. Cy3 shows brighter red fluorescence.

The assay is based on the observation that soon after initiating apoptosis, cells translocate the membrane phospholipid phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface. Once on the cell surface, PS can easily be detected by staining with a fluorescent conjugate of Annexin V, a protein that has a strong natural affinity for PS. The one-step staining procedure takes only 10 minutes. In addition, the assay can be directly performed on live cells, without the need of fixation. The Annexin V-Cy3 Apoptosis Detection Kit Plus includes annexin V-Cy3, SYTOX green dye, and binding buffer. The SYTOX green dye is impermeant to live cells and apoptotic cells, but stains necrotic cells with intense green fluorescence by binding to cellular nucleic acids. After staining a cell population with annexin V-Cy3 and SYTOX Green dye in the provided binding buffer, apoptotic cells show red fluorescence, dead cells show green fluorescence and live cells show little or no fluorescence. These populations can easily be distinguished by Fluorescence microscopy using FITC and rhodamine filters or by flow cytometry using the FL1 channel (Ex. 488 nm/Em. 530 nm) for SYTOX Green dye and FL2 channel for Annexin V-Cy3 (Ex. 543 nm/Em. 570 nm).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty