Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Calpain 1 Inhibitor Screening Kit, BioVision (Fluorometric)

Supplier: BioVision
AB283373-100TEST
10186-552EA 1461 USD InStock
10186-552
Human Calpain 1 Inhibitor Screening Kit, BioVision (Fluorometric)
Assays Cellular Assays
HTS, includes Calpain 1 inhibitor

Detection method- Fluorescence (Ex/Em= 400/505 nm).
Species reactivity- Mammalian
Applications- The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput screening of Calpain 1 inhibitors. Inhibitor control (Calpain inhibitor 1, ALLN) is included to compare the efficacy of test inhibitors.

Calpain 1 is calcium-dependent, non-lysosomal cysteine protease with low calcium requirement. It is expressed ubiquitously and in the brain, mainly localized to the cell body and dendrites of neurons and to a lesser extent in axons and glial cells. Hyperactivation of calpains is implicated in a number of pathologies associated with altered calcium homeostasis such as Alzheimer’s disease as well as secondary degeneration resulting from acute cellular stress following cerebral (neuronal) ischemia, traumatic brain injury and spinal cord injury. BioVision’s human calpain 1 inhibitor screening kit is based on the detection of cleavage of calpain substrate Ac-LLY-AFC. Ac-LLY-AFC possess very weak fluorescence; upon cleavage of the substrate by calpain 1, free AFC can be quantified fluorometrically (Ex/Em = 400/505 nm). In the presence of potent Calpain 1 inhibitor, the hydrolyzation of substrate will be impeded. The kit provides a rapid, simple, sensitive, and reliable test suitable for high throughput screening of Calpain 1 inhibitors. Inhibitor control (Calpain inhibitor 1, ALLN) is included to compare the efficacy of test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty