Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric Oxide Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB65327-200
10186-558EA 1055.91 USD InStock
10186-558
Nitric Oxide Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Detects total nitrate/nitrite

Detection method- Fluorescence (Ex/Em 380/461 nm)
Species reactivity- Mammalian
Applications- Measurement of total nitrate/nitrite concentration

Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. Since NO is rapidly converted to nitrite (NO²ˉ) and nitrate (NOᶟˉ), the total concentration of nitrite and nitrate is used as a quantitative measure of NO production. BioVision’s Nitric Oxide Fluorometric Assay Kit provides an accurate and convenient measurement of total nitrate/nitrite concentration in a simple two-step process. In the first step nitrate is converted to nitrite by nitrate reductase. In the second step, nitrite reacts with the fluorescent probe DAN (2, 3 diaminonaphthalene). NaOH enhances the fluorescent yield. The fluorescent intensity is proportional to the total nitric oxide production. The kit has been tested with culture media, plasma, and tissue homogenates.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty