Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ADP/ATP Ratio Bioluminescence Assay Kit, ApoSENSOR, BioVision

Supplier: BioVision
AB65313-200
10186-562EA 809.84 USD InStock
10186-562
ADP/ATP Ratio Bioluminescence Assay Kit, ApoSENSOR, BioVision
Assays Cellular Assays
Sensitive Assay, HTS

Detection method- Luminometer or Beta Counter.
Species reactivity- Mammalian
Applications- Bioluminescent detection of the ADP and ATP levels for a rapid screening of apoptosis, necrosis, growth arrest, and cell proliferation simultaneously in mammalian cells.

The changes in ADP/ATP ratio have been used to differentiate the different modes of cell death and viability. Increased levels of ATP and decreased levels of ADP have been recognized in proliferating cells. In contrast, decreased levels of ATP and increased levels of ADP are recognized in apoptotic cells. The decrease in ATP and increase in ADP are much more pronounced in necrosis than apoptosis. The ApoSENSORTM ADP/ATP Ratio Assay kit utilizes bioluminescent detection of the ADP and ATP levels for a rapid screening of apoptosis, necrosis, growth arrest, and cell proliferation simultaneously in mammalian cells. The assay utilizes the enzyme luciferase to catalyze the formation of light from ATP and luciferin, and the light can be measured using a luminometer or Beta Counter. ADP level is measured by its conversion to ATP that is subsequently detected using the same reaction. The assay can be fully automatic for high throughput and is highly sensitive (detects 100 mammalian cells/well).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty