Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mitochondria/Cytosol Fractionation Kit, BioVision

Supplier: BioVision
AB65320-25 AB65320-100
10186-564EA 959.25 USD InStock
10186-564 10186-566
Mitochondria/Cytosol Fractionation Kit, BioVision
Assays Cellular Assays
Isolates mitochondria/cytosol w/o ultracentrifugation

Detection method- Western blotting, ELISA, or other assays.
Species reactivity- Mammalian
Kit size- 25, 100 assays
Applications- Effective isolation of a highly enriched mitochondrial fraction from cytosolic fraction of mammalian cells including both apoptotic and nonapoptotic cells.

The Mitochondria/Cytosol Fractionation Kit provides unique formulations of reagents for effective isolation of a highly enriched mitochondrial fraction from cytosolic fraction of mammalian cells including both apoptotic and nonapoptotic cells. The enriched mitochondrial and cytosolic fractions can be used for studying apoptotic and signal transduction pathways to detect translocation of factors interested between the two fractions by Western blotting, ELISA, or other assays. Procedures are simple and easy to perform, no ultracentrifugations and toxic chemicals are involved.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty