Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GST Fluorometric Activity Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB65325-100
10186-574EA 504.95 USD InStock
10186-574
GST Fluorometric Activity Assay Kit, BioVision
Assays Cellular Assays
Highly sensitive in vitro assay

Detection method- Fluorescence (Ex/Em 380/461 nm)
Species reactivity- Mammalian
Applications- GST level in samples can be easily detected using a fluorometer or a 96-well fluorometric plate reader. The kit can detect GST activity in crude cell lysate or purified protein fraction, and also quantitate GST-tagged fusion protein. Detects < 0.5 mU.

Glutathione S-transferase (GST) is a family of enzymes that play an important role in detoxification of xenobiotics. GST catalyzes the formation of the thiol group of glutathione to electrophilic xenobiotics. It utilizes glutathione to scavenge potentially toxic compounds including those produced as a result of oxidative stress and is part of the defense mechanism against the mutagenic, carcinogenic and toxic effects of such compounds. The GST Fluorometric Activity Assay Kit provides a simple, fluorescence-based in vitro assay for detecting the GST activity using fluorescence plate reader. The assay utilizes monochlorobimane (MCB), a dye that reacts with glutathione. The free form of MCB is almost nonfluorescent, whereas the dye fluoresces blue (ex/em = 380/461 nm) when reacts with glutathione. GST catalyzes the MCB-glutathione reactions and the fluorescence levels are proportionally to the amounts of GST presence in the reaction. Thus, the GST level in samples can be easily detected using a fluorometer or a 96-well fluorometric plate reader. The kit can detect GST activity in crude cell lysate or purified protein fraction, and also quantitate GST-tagged fusion protein. Detects < 0.5 mU.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty