Would you like to log in again?
Genomic DNA Isolation Kit, BioVision, Inc.
Supplier: BioVision
AB65358-50
76200-088EA 268.85 USD InStock
76200-088
Genomic DNA Isolation Kit, BioVision, Inc.
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The kit provides a simple and convenient procedure for rapid isolation of genomic DNA from mammalian cells and tissue samples with high yield and purity. The novel method requires less than 90 minutes to prepare highly pure genomic DNA. The extracted genomic DNA is free from protein and RNA, and suitable for a variety of applications such as PCR, DNA hybridization, enzyme manipulation, cloning, Southern blot, and array-based experiments.
  • Isolates highly pure genomic DNA within 90 min.
Simple procedure, takes less than 2 hours
Fast and convenient
Provides unique formulations of buffers and reagents for isolation of highly pure genomic DNA
Caution: Storage −20 °C (stable for 12 months)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty