Would you like to log in again?
Mitochondrial Protein IP Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB239710-50TEST
10186-600EA 834.77 USD InStock
10186-600
Mitochondrial Protein IP Kit, BioVision
Assays Cellular Assays
For IP/Co-IP/Pull-down of mt
Application
Optimized for compatibility with immunoprecipitation (IP and co-IP) and pull-down using tagged proteins.
Gentle formulation for maintenance of stable mitochondrial complexes.
Compatible with SDS PAGE, 2D gel, Blue-Native gel, Mass spectrometry.
Functional Assays and enzymatic assays.
Mitochondria are the power house of the cells and play an essential role in energy production. Damage to the mitochondria activates signaling pathways that induce apoptosis. Mitochondria also regulate and mediate transport of the metabolites and ions needed for oxidative phosphorylation and maintenance of membrane potential for ATP synthesis. Mitochondrial dysfunction leads to several disorders like cardiac dysfunction, diabetes, aging and neurological disorder, mainly caused by mutations in mitochondrial DNA or in nuclear genes that code for mitochondrial components. Thus, mitochondria have several different functions in the cell.
BioVision ready to use mitochondria Protein IP Buffer is optimized for immunoprecipitation (IP and co-IP) using mitochondria and mitochondrial extracts. The buffer is a gentle formulation, which maintains the stability of mitochondrial complexes. The Mitochondrial Protein IP kit is provided with different choices of detergents like n-Dodecyl-beta-D-maltoside, Triton X-100 and digitonin to achieve different stringency conditions for protein-protein interaction studies. Triton X-100 is the most commonly used detergent especially for membrane protein solubilization. However, in case of fragile complexes digitonin or n-Dodecyl-beta-D-maltoside is the choice of detergents.
proteins or complexes
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty