Would you like to log in again?
Mitochondria Isolation Kit for Tissue and Cultured Cells, BioVision
Supplier: BioVision
AB288084-50TEST
10186-602EA 759.21 USD InStock
10186-602
Mitochondria Isolation Kit for Tissue and Cultured Cells, BioVision
Assays Cellular Assays
Isolates highly pure, intact & functional mitochondria
Useful for mitochondrial respiration studies, assembly of the complexes, apoptosis, mtDNA and mtRNA, and protein profiling.
Western blot and ELISA.
Mitochondria are the powerhouse of the cells because they generate most of the supply of energy in the form of adenosine tri-phosphate (ATP). Mitochondria are double membrane organelles: an outer membrane and a folded inner membrane called cristae. Isolated mitochondria are the sample of choice to study mitochondrial respiration, assembly of the respiratory complexes, apoptosis, mtDNA and mtRNA, and mitochondrial protein profiling. The kit offers two options for the isolation of intact mitochondria. The first option is utilizing a reagent-based method allowing parallel processing of multiple samples. The second option uses traditional dounce homogenization, which provides better mitochondrial yield.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty