Would you like to log in again?
Quick Cell Proliferation Colorimetric Assay Kit, BioVision
Supplier: BioVision
K301-2500 AB65473-500
10186-446EA 1247.84 USD InStock
10186-446 10186-448
Quick Cell Proliferation Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Sensitive, Non-radioactive Assay
Detection- Absorbance (420-480 nm)
Kit size- 500, 2500
Applications- This fast and sensitive cell proliferation assay uses a water-soluble tetrazolium salt WST-1. The assay measures cell proliferation in response to growth factors/cytokines/mitogens/and nutrients etc.
The Quick Cell Proliferation Assay Kit provides all reagents and detailed instructions for a fast and sensitive quantification of cell proliferation and viability. The assay is based on the cleavage of the tetrazolium salt WST-1 to formazan by cellular mitochondrial dehydrogenases. Expansion in the number of viable cells resulted in an increase in the activity of the mitochondrial dehydrogenases, which leads to the increase in the amount of formazan dye formed. The formazan dye produced by viable cells can be quantified by multi-well spectrophotometer (microtiter plate reader) by measuring the absorbance of the dye solution at 440 nm. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds. The new method is so simple, requiring no washing, no harvesting, and no solubilization steps, and is faster and more sensitive than MTT, XTT, or MTS-based assays. The entire assay can be performed in a microtiter plate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty