Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VisionBlue™ Quick Cell Viability Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB102501-500TEST AB102501-2500TEST
10186-608EA 833.77 USD InStock
10186-608 10186-610
VisionBlue™ Quick Cell Viability Fluorometric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive cell viability kit

Detection method- Fluorescence (Ex/Em 530–570/ 590-620 nm)
Species reactivity- Mammalian
Kit size- Convenient sizes (500, 2500)
Applications- This fluorescence-based single-step cell viabilty assay utilizes the redox dye (Resazurin) which is not fluorescent, but upon reduction by metabolically active cells, it becomes highly fluorescent (Ex = 530-570 nm; Em = 590 -620 nm). Therefore, viable metabolically active cells can be easily measured.

The VisionBlue™ Quick Cell Proliferation Assay Kit provides by far the most sensitive and easiest means for quantifying cell proliferation and viability. Simply add the single reagent to cell culture, incubate and read the fluorescence intensity. The VisionBlue™ assay utilizes the redox dye resazurin which is not fluorescent, but upon reduction by viable metabolically active cells, the dye becomes highly fluorescent (Ex = 530-570 nm; Em = 590-620 nm). Therefore, viable metabolically active cells can be easily measured.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty