Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LDH-Cytotoxicity Colorimetric Assay Kit, BioVision

Supplier: BioVision
AB65391-400
10186-614EA 636.05 USD InStock
10186-614
LDH-Cytotoxicity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (500 nm)
Sample type- Cell culture (adherent and suspension)
Species reactivity- Mammalian
Applications- This assay provides a convenient and non-radioactive alternative to the [3H]-thymidine and the [51Cr] release assays The assay is based on measurement of activity of lactate dehydrogenase (LDH) which is a stable enzyme normally found in the cytosol of all cells but rapidly releases into the supernatant upon damage of plasma membrane.

Cell death or cytotoxicity is classically evaluated by the quantification of plasma membrane damage. The LDH-Cytotoxicity Assay Kit provides a fast and simple method for quantitating cytotoxicity based on the measurement of activity of lactate dehydrogenase (LDH) released from damaged cells. Unlike many other cytoplasmic enzymes which exist in many cells either in low amount (e.g., alkaline and acid phosphatase) or unstable, LDH is a stable cytoplasmic enzyme present in all cells and rapidly released into the cell culture supernatant upon damage of the plasma membrane. LDH activity can be determined by a coupled enzymatic reaction: LDH oxidizes lactate to pyruvate which then reacts with tetrazolium salt INT to form formazan. The increase in the amount of formazan produced in culture supernatant directly correlates to the increase in the number of lysed cells. The formazan dye is water-soluble and can be detected by spectrophotometer at 500 nm. The LDH-cytotoxicity assay is sensitive, convenient, and precise, and is applicable to a variety of cytotoxicity studies. Assay takes ~0.5–1 hr.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty