Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citrate Synthase Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB239712-100TEST
10186-620EA 979.56 USD InStock
10186-620
Citrate Synthase Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Detection method- Absorbance (412 nm)
Applications- - Measurement of citrate synthase activity in various tissues/cells
- Analysis of intact mitochondria

Citrate Synthase (EC 2.3.3.1) is a key enzyme that is present in all living organisms. It catalyzes the conversion of acetyl-CoA and oxaloacetate into citrate and serves as a marker for intact mitochondria. A recent study showed that increased activity of mitochondrial Citrate Synthase is directly associated with metabolic and endocrine abnormalities such as obesity. In BioVision’s Citrate Synthase Activity Assay Kit, Citrate Synthase reacts with substrate mix to form an intermediate, which subsequently reacts with developer to generate the colored product. The rate of color development is proportional to the enzyme activity. The assay is simple, rapid and can detect Citrate Synthase activity less than 1 mU in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty