Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SIRT2 Inhibitor Screening Assay Kit (Fluorometric), BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB283375-100TEST
10186-624EA 1133.8 USD InStock
10186-624
SIRT2 Inhibitor Screening Assay Kit (Fluorometric), BioVision
Assays Cellular Assays
HTS, includes inhibitor

Detection method- Fluorescence (Ex/Em 395/541 nm)
Species reactivity- Mammalian
Application- Screening and characterization of Sirtuin inhibitors.

Sirtuin or Sir2 proteins are a class of proteins that possess either histone deacetylase or mono-ribosyltransferase activity. Sirtuins have been implicated in influencing aging and regulating transcription, apoptosis and stress resistance, as well as energy efficiency and alertness during low-calorie situations. Unlike other known protein deacetylases, which simply hydrolyze acetyl-lysine residues, the sirtuin-mediated deacetylation reaction couples lysine deacetylation to NAD hydrolysis. This hydrolysis yields O-acetyl-ADP-ribose, the deacetylated substrate and nicotinamide, itself an inhibitor of sirtuin activity. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. In BioVision’s Sirtuin inhibitor screening Kit, Sirtuin deacetylates the substrate and then the developer cleaves the deacetylated substrate to release the fluorescent group, which can be detected fluorometrically at Ex/Em = 395/541 nm. In the presence of SIRT inhibitor, the deacetylation will be impeded which prevents the cleavage of the substrate to release the fluorescent group. The kit provides a rapid, simple, sensitive, and reliable test, which is also suitable for high-throughput screening of SIRT2 inhibitors. Inhibitor control (Nicotinamide) is included to compare the efficacy of the test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty