Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HDAC Activity Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB1438-96
10186-626EA 529.95 USD InStock
10186-626
HDAC Activity Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Two step assay, HTS

Detection method- Fluorescence (Ex/Em 350-380/440-460 nm)
Species reactivity- Mammalian
Applications- Well suited for high throughput applications. HDAC inhibitors and substrates and related antibodies are also available separately.

Inhibition of histone deacetylase (HDAC) has been implicated to modulate transcription and induce apoptosis or differentiation in cancer cells. However, screening compounds for HDAC inhibition has been difficult due to the lack of convenient tools for analyzing HDAC activity. The Fluorometric HDAC Activity Assay Kit provides a fast and fluorescence-based method that eliminates radioactivity, extractions, or chromatography, as used in traditional assays. The new procedure requires only two easy steps, both performed on the same microtiter plate. First, the HDAC substrate, which comprises an acetylated lysine side chain, is incubated with a sample containing HDAC activity (e.g., HeLa nuclear extract). Deacetylation of the substrate sensitizes the substrate, so that further treatment with the Lysine Developer produces a fluorophore. The fluorophore can be easily analyzed using a fluorescence plate reader or a fluorometer. The assay is well suited for high throughput screening applications. HDAC inhibitors and antibodies are also available separately.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty