Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HAT Activity Colorimetric Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB65352-100
10186-630EA 645 USD InStock
10186-630
HAT Activity Colorimetric Assay Kit, BioVision
Assays Cellular Assays
Non-radioactive assay for end point & kinetic studies

Detection method- Absorbance (440 nm)
Species reactivity- Mammalian
Applications- Detects HAT activity by colorimetric method in mammalian samples

Histone acetyltransferases (HATs) have been implicated to play a crucial role in various cellular functions, such as gene transcription, differentiation, and proliferation. BioVision’s HAT Activity Colorimetric Assay Kit offers a convenient, nonradioactive system for a rapid and sensitive detection of HAT activity in mammalian samples. The kit utilizes active Nuclear Extract (NE) as a positive control and acetyl-CoA as a cofactor. Acetylation of peptide substrate by active HAT releases the free form of CoA which then serves as an essential coenzyme for producing NADH. NADH can easily be detected spectrophotometrically upon reacting with a soluble tetrazolium dye. The detection can be continuous and suitable for kinetic studies. The kit provides a simple, straightforward protocol for a complete assay.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty