Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NAD/NADH Quantitation Colorimetric Kit, BioVision

Supplier: BioVision
K337-100
10186-636EA 1081.93 USD InStock
10186-636
NAD/NADH Quantitation Colorimetric Kit, BioVision
Assays Cellular Assays
No NADP or NADPH Detection

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Applications- BioVision’s NADH/NAD Quantification Kit provides a convenient tool for sensitive detection of the intracellular nucleotides: NADH, NAD and their ratio.

Assay of nicotinamide nucleotides is of continual interest in the studies of energy transforming and redox state of cells or tissues. BioVision’s NADH/NAD Quantification Kit provides a convenient tool for sensitive detection of the intracellular nucleotides: NADH, NAD and their ratio. The NAD Cycling Enzyme Mix in the kit specifically recognizes NADH/NAD in an enzyme cycling reaction. There is no requirement to purify NADH/NAD from samples. The reaction specifically detects NADH and NAD, but not NADP nor NADPH. The enzyme cycling reaction significantly increases the detection sensitivity and specificity. NADt (NAD and NADH) or NADH can be easily quantified by comparing with standard NADH.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty