Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HDAC Inhibitor Drug Screening Kit (Fluorometric), BioVision

Supplier: BioVision
AB283378-100TEST
10186-640EA 862.91 USD InStock
10186-640
HDAC Inhibitor Drug Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Non-radioactive, Two Easy Steps, HTS

Detection method- Fluorescence (Ex/Em 350-380/440-460 nm)
Species reactivity- Mammalian
Applications- Screening of compounds for HDAC inhibition by analyzing HDAC activity

Inhibition of histone deacetylase (HDAC) has been implicated to modulate transcription and to induce apoptosis or differentiation in cancer cells. However, screening of compounds for HDAC inhibition has been difficult due to the lack of convenient tools for analyzing HDAC activity. The new HDAC Inhibitor Drug Screening Kit provides a fast, fluorescence-based method that eliminates radioactivity, extractions, or chromatography, as used in traditional assays. The new procedure requires only two easy steps, both performed on the same microtiter plate. First, your inhibitor candidates are mixed with HeLa Nuclear Extract and HDAC fluorometric substrate, which comprises an acetylated lysine side chain. Deacetylation of the substrate sensitizes the substrate, so that, in the second step, treatment with the Lysine Developer produces a fluorophore. The fluorophore can be easily analyzed using a fluorescence plate reader or a fluorometer. The assay is well suited for high throughput screening applications.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty