Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HDAC2 Immunoprecipitation (IP) and Activity Assay Kit, BioVision

Supplier: BioVision
AB287881-25TEST
10186-642EA 1333 USD InStock
10186-642
HDAC2 Immunoprecipitation (IP) and Activity Assay Kit, BioVision
Assays Cellular Assays
Only kit on the market for both IP as well as measuring activity of HDAC2

Detection method - Fluorescence (Ex/Em=380/500 nm)
Applications:
- Immunoprecipitation of HDAC2 complex from cell, nuclear, or tissue extract(s)
- Measurement of HDAC2 activity of immunoprecipitated complex/purified enzyme
- Screening for activators or inhibitors of HDAC2

Histone deacetylases (HDACs) play a central role in controlling cell cycle regulation, cell differentiation, and tissue development. These proteins have crucial roles in development and physiology, especially in the heart and nervous system. It is also deeply involved in cellular proliferation, cell cycle and apoptosis. The HDAC2 IP & Activity Assay Kit provides an antibody-based method to specifically immunoprecipitate the HDAC2 complex from cells & tissues and to measure HDAC2 activity fluorometrically. HDAC2 is immunoprecipitated from cell or nuclear extract(s) using HDAC2 antibody followed by capturing the complex on protein-A/G beads. The immunoprecipitated complex is incubated with the HDAC substrate. Only the deacetylated substrate is cleaved by the Developer to produce a fluorophore, which can be easily analyzed using a fluorescence plate reader.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty