Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HDAC1 Immunoprecipitation (IP) and Activity Assay Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB287873-25TEST
10186-644EA 1317.68 USD InStock
10186-644
HDAC1 Immunoprecipitation (IP) and Activity Assay Kit, BioVision
Assays Cellular Assays
Only kit on the market for both IP and measuring activity of HDAC1

Detection method - Fluorescence (Ex/Em=380/500 nm)
Applications:
- Immunoprecipitation of HDAC1 complex from cell, nuclear, or tissue extract(s)
- Measurement of HDAC1 activity of immunoprecipitated complex/purified enzyme
- Screening for activators or inhibitors of HDAC1

Histone deacetylases (HDACs) play a central role in controlling cell cycle regulation, cell differentiation, and tissue development. HDAC1 is ubiquitously expressed and plays a role in cellular senescence, cardiomyocyte differentiation and hypertrophy, aging of the liver, myelination, and adult neurogenesis to name a few functions. The HDAC1 IP & Activity Assay Kit provides an antibody-based method to specifically immunoprecipitate the HDAC1 complex from cells & tissues and to measure HDAC1 activity fluorometrically. HDAC1 is immunoprecipitated from cell, nuclear or tissue extract(s) using HDAC1 antibody followed by capturing the complex on protein-A/G beads. The immunoprecipitated complex is incubated with the HDAC substrate. Only the deacetylated substrate is cleaved by the Developer to produce a fluorophore, which can be easily analyzed using a fluorescence plate reader.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty