Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PicoProbe™ NADPH Quantitation Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB287848-100TEST
10186-650EA 994.41 USD InStock
10186-650
PicoProbe™ NADPH Quantitation Fluorometric Assay Kit, BioVision
Assays Cellular Assays
The most sensitive kit on the market

Detection method- Fluorescence (Ex/Em = 535/587 nm)
Applications:
- Measurement of NADPH in various tissues/cells
- Analysis of metabolism and cell signaling in various cells

NADP (nicotinamide dinucleotide) is a major electron donor for anabolic reactions such as lipid and nucleic acid synthesis. The reduced form, NADPH is a cofactor for many redox enzymes. It is of increasing interest to be able to measure low level of intracellular NADPH. BioVision’s PicoProbe™ NADPH Quantitation Assay kit provides a highly sensitive method to detect intracellular NADPH. NADH is not detected as the NADPH Recycling Enzyme Mix specifically recognizes NADPH in the enzyme recycling reaction. The assay is rapid, simple and does not require purifying NADPH from samples. The kit can measure less than 40 nM NADPH in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty