Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ATP Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
K354-100
10186-652EA 1030.65 USD InStock
10186-652
ATP Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Sensitive, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- All
Applications- The assay can detect as low as 50 picomol (1 µM) of ATP in various samples.

ATP is the primary energy currency of living systems. Virtually all energy requiring processes utilize the chemical energy stored in the phosphate bond of ATP. ATP is formed exclusively in the mitochondria and a variety of genetic diseases can affect ATP formation in the mitochondria. There are a number of commercially available ATP assays which detects femtomoles or less of ATP by measuring luminescence (BioVision Kit 254-200, for example) but these kits require specialized luminescence instrumentation and utilize luciferase which can be difficult to maintain in active form. BioVision newly developed ATP Colorimetric and Fluorometric Assay kit is designed to be a robust, simple method which utilizes the phosphorylation of glycerol to generate a product that is easily quantified by colorimetric (λmax = 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The assay can detect as low as 50 picomol (1 µM) of ATP in various samples. The kit provides sufficient reagents for 100 assays.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty