Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ADP Colorimetric/Fluorometric Assay Kit, BioVision

Supplier: BioVision
AB83359-100
10186-654EA 622.04 USD InStock
10186-654
ADP Colorimetric/Fluorometric Assay Kit, BioVision
Assays Cellular Assays
Highly Stable, HTS

Detection method- Absorbance (570 nm) or Fluorescence (Ex/Em 535/587 nm)
Species reactivity- Mammalian
Applications- In the assay, ADP is converted to ATP and pyruvate. The assay can detect as low as 1 µM ADP in biological samples.

ADP is a product of ATP dephosphorylation and it can be rephosphorylated to ATP. De-phosphorylation and rephosphorylation occur via various phosphatases, phosphorylases and kinases. ADP is stored in platelets and can be released to interact with a variety of purinergic receptors. ADP levels regulate several enzymes involved in intermediary metabolism. ADP conversion to ATP primarily occurs within the mitochondrion and chloroplast although several such processes occur in the cytoplasm. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, the luciferase system is unstable and luminescence equipment is not generally available in most laboratories. BioVision’s newly designed ADP Assay Kit provides a convenient colorimetric and fluorometric means to measure ADP level. In the assay, ADP is converted to ATP and pyruvate. The generated pyruvate can be quantified by colorimetric (λmax = 570 nm) or fluorometric method (Ex/Em 535/587 nm). The assay is simple, sensitive, stable and high-throughput adaptable. The assay can detect as low as 1 µM ADP in biological samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty